Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005376.2 | NP_001005376.1 | 3 | 467 | ||
NM_001005377.2 | NP_001005377.1 | 3 | 467 | |||
NM_001301037.1 | NP_001287966.1 | 3 | 467 | |||
NM_002659.3 | NP_002650.1 | 3 | 467 | |||
XM_005258990.4 | XP_005259047.1 | 3 | 289 | |||
XM_011527028.2 | XP_011525330.1 | 3 | 289 | |||
XM_011527029.1 | XP_011525331.1 | 3 | 289 | |||
XM_011527030.2 | XP_011525332.1 | 3 | 289 | |||
XM_011527031.2 | XP_011525333.1 | 3 | 289 | |||
XM_017026872.1 | XP_016882361.1 | 3 | 702 | |||
GenBank mRNA | A18757.1 | - | 3 | 287 | ||
AF257789.1 | - | 3 | 241 | |||
AK290774.1 | - | 3 | 287 | |||
AY029180.1 | - | 3 | 287 | |||
BC002788.2 | - | 3 | 306 | 3617894 | ||
CA495321.1 | - | 3 | 467 | |||
CF126152.1 | - | 3 | 274 | |||
CR456952.1 | - | 3 | 241 | |||
M83246.1 | - | 3 | 258 | |||
U08839.1 | - | 3 | 667 | |||
X51675.1 | - | 3 | 287 | |||
X74039.1 | - | 3 | 287 |