Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135685.1 | NP_001129157.1 | 17, 18 | 2145 | ||
NM_002344.5 | NP_002335.2 | 19, 20 | 2535 | |||
NM_206961.3 | NP_996844.1 | 18, 19 | 2352 | |||
XM_011521556.2 | XP_011519858.1 | 19, 20 | 2583 | |||
XM_011521557.2 | XP_011519859.1 | 18, 19 | 2415 | |||
XM_017022181.1 | XP_016877670.1 | 19, 20 | 2598 | |||
XM_017022182.1 | XP_016877671.1 | 18, 19 | 2454 | |||
XM_017022183.1 | XP_016877672.1 | 18, 19 | 2343 | |||
GenBank mRNA | AK296892.1 | - | 17, 18 | 2145 | ||
AK314006.1 | - | 19, 20 | 2502 | |||
BC045607.1 | - | 17, 18 | 2852 | 5270700 | ||
BC143903.1 | - | 13, 14 | 1538 | |||
D16105.1 | - | 19, 20 | 2454 | |||
X52213.1 | - | 12, 13 | 1415 | |||
X60702.1 | - | 18, 19 | 2344 |