Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018137.2 | NP_001018147.1 | 4 | 540 | ||
NM_001018138.1 | NP_001018148.1 | 3 | 398 | |||
NM_001018139.2 | NP_001018149.1 | 4 | 356 | |||
NM_002512.3 | NP_002503.1 | 4 | 519 | |||
GenBank mRNA | BC002476.2 | - | 4 | 321 | 3349768 | |
BC095458.1 | - | 4 | 330 | 30524221 | ||
BF034471.1 | - | 4 | 441 | |||
BF058708.1 | - | 3 | 398 | |||
BM556410.1 | - | 4 | 356 | |||
BT007045.1 | - | 3 | 289 | |||
CR541718.1 | - | 3 | 289 | |||
EU794639.1 | - | 4 | 338 | |||
FJ655905.1 | - | 162 | ||||
L16785.1 | - | 4 | 338 | |||
M36981.1 | - | 4 | 361 | |||
X58965.1 | - | 4 | 361 |
Gene Symbol: NME1-NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018136.2 | NP_001018146.1 | 7 | 765 | ||
NR_037149.1 | 8 | 985 | ||||
GenBank mRNA | AK313448.1 | - | 7 | 722 | ||
BC107894.1 | - | 8 | 887 | 6717551 | ||
BC133029.1 | - | 7 | 668 | 40147051 | ||
BC133031.1 | - | 7 | 668 | 40147053 | ||
BG388883.1 | - | 692 | ||||
DQ109675.1 | - | 7 | 685 |