Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002970.3 | NP_002961.1 | 6 | 829 | ||
NR_027783.2 | 7 | 939 | ||||
XM_017029720.1 | XP_016885209.1 | 3 | 1457 | |||
GenBank mRNA | AF251292.1 | - | 6 | 848 | ||
AL050290.1 | - | 7 | 900 | |||
BC002503.2 | - | 6 | 790 | 3051095 | ||
BC008424.1 | - | 6 | 814 | 4134501 | ||
M55580.1 | - | 640 | ||||
M77693.1 | - | 6 | 800 |