Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003032.2 | NP_003023.1 | 7 | 4176 | ||
NM_173216.2 | NP_775323.1 | 8 | 4499 | |||
NM_173217.2 | NP_775324.1 | 7 | 3842 | |||
XM_005247719.1 | XP_005247776.1 | 7 | 4087 | |||
XM_005247720.1 | XP_005247777.1 | 7 | 4145 | |||
XM_006713734.1 | XP_006713797.1 | 9 | 4754 | |||
XM_011513085.1 | XP_011511387.1 | 9 | 4762 | |||
XM_011513086.1 | XP_011511388.1 | 7 | 4145 | |||
XM_017007065.1 | XP_016862554.1 | 10 | 4846 | |||
XM_017007066.1 | XP_016862555.1 | 7 | 4174 | |||
XM_017007067.1 | XP_016862556.1 | 7 | 4294 | |||
XM_017007068.1 | XP_016862557.1 | 7 | 4179 | |||
GenBank mRNA | AF007133.1 | - | 1 | 2020 | ||
AK128726.1 | - | 3 | 4790 | |||
BC031476.1 | - | 7 | 4170 | 4339124 | ||
BC040009.1 | - | 7 | 4176 | 5557868 |