Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079880.1 | NP_001073349.1 | 19 | 3105 | ||
NM_001079881.1 | NP_001073350.1 | 19 | 2969 | |||
NM_001079882.1 | NP_001073351.1 | 17 | 2706 | |||
NM_016457.4 | NP_057541.2 | 18 | 3384 | |||
XM_005258716.2 | XP_005258773.2 | 17 | 2362 | |||
GenBank mRNA | AF151021.1 | - | 6 | 910 | ||
AF309082.1 | - | 18 | 2666 | |||
AK074673.1 | - | 17 | 2345 | |||
AK075266.1 | - | 17 | 2455 | |||
AK092896.1 | - | 18 | 2756 | |||
AK095884.1 | - | 19 | 2648 | |||
AK300339.1 | - | 19 | 2949 | |||
AK303629.1 | - | 17 | 2360 | |||
AL050147.1 | - | 12 | 1621 | |||
BC015472.1 | - | 9 | 1083 | |||
BC025307.1 | - | 14 | 1955 |