Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320611.1 | NP_001307540.1 | 3, 4 | 841 | ||
NM_002266.3 | NP_002257.1 | 3, 4 | 483 | |||
GenBank mRNA | AK024833.1 | - | 3, 4 | 351 | ||
AK291954.1 | - | 3, 4 | 350 | |||
BC005978.1 | - | 3, 4 | 350 | 4105058 | ||
BC013257.1 | - | 3, 4 | 346 | |||
BC034443.1 | - | 3, 4 | 309 | |||
BC053343.1 | - | 3, 4 | 339 | 4450470 | ||
BC067848.1 | - | 3, 4 | 582 | 4825235 | ||
BC146905.1 | - | 3, 4 | 337 | 9056833 | ||
BT006665.1 | - | 2, 3 | 221 | |||
DC388846.1 | - | 3, 4 | 483 | |||
U09559.1 | - | 3, 4 | 353 | |||
U28386.1 | - | 3, 4 | 353 |