Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKACB SIRNA Assays ›
Gene Symbol:
PRKACB
Gene Name:
protein kinase cAMP-activated catalytic subunit beta
Gene Aliases:
PKA C-beta, PKACB
Chromosome Location:
Chr.1: 84077975 - 84238498 on Build GRCh38
UniGene ID:
Hs.487325
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
111266
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKACB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242857.2
NP_001229786.1
6
451
NM_001242858.2
NP_001229787.1
3
394
NM_001242859.2
NP_001229788.1
5
442
NM_001242860.2
NP_001229789.1
6
586
NM_001242861.2
NP_001229790.1
5
577
NM_001242862.2
NP_001229791.1
2
190
NM_001300915.1
NP_001287844.1
6
586
NM_001300916.1
NP_001287845.1
3
456
NM_001300917.1
NP_001287846.1
3
394
NM_002731.3
NP_002722.1
3
552
NM_182948.3
NP_891993.1
3
456
NM_207578.2
NP_997461.1
3
552
XM_005271015.3
XP_005271072.1
5
403
XM_005271016.1
XP_005271073.1
5
528
XM_005271017.3
XP_005271074.1
4
393
XM_005271018.4
XP_005271075.1
4
713
XM_005271019.1
XP_005271076.1
3
469
XM_005271020.1
XP_005271077.1
3
436
XM_006710758.1
XP_006710821.1
5
542
XM_017001706.1
XP_016857195.1
6
446
XM_017001707.1
XP_016857196.1
6
446
XM_017001708.1
XP_016857197.1
6
547
XM_017001709.1
XP_016857198.1
5
244
XM_017001710.1
XP_016857199.1
5
542
XM_017001711.1
XP_016857200.1
5
244
XM_017001712.1
XP_016857201.1
5
542
XM_017001713.1
XP_016857202.1
3
494
XM_017001714.1
XP_016857203.1
5
528
XM_017001715.1
XP_016857204.1
5
528
XM_017001716.1
XP_016857205.1
4
396
XM_017001717.1
XP_016857206.1
4
717
XM_017001718.1
XP_016857207.1
3
371
XM_017001719.1
XP_016857208.1
3
469
XM_017001720.1
XP_016857209.1
3
469
GenBank mRNA
AB209189.1
-
6
522
AB451239.1
-
3
201
AB451364.1
-
3
201
AK091420.1
-
5
251
AK296482.1
-
6
253
AK304322.1
-
3
381
AK304375.1
-
3
196
AK314550.1
-
3
448
AK316115.1
-
2
163
BC016285.1
-
3
383
3916100
BC035058.1
-
3
470
5262372
BI598037.1
-
3
266
BM905428.1
-
3
338
BX485407.1
-
3
437
BX537705.1
-
3
433
BX641026.1
-
3
432
BX647515.1
-
6
531
CR936631.1
-
5
245
DA228647.1
-
2
191
DC322547.1
-
6
452
DC418574.1
-
3
456
M34181.1
-
3
248
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
stimulatory C-type lectin receptor signaling pathway
renal water homeostasis
protein phosphorylation
signal transduction
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
blood coagulation
activation of protein kinase A activity
lipoprotein metabolic process
negative regulation of meiotic cell cycle
regulation of protein processing
cellular response to glucagon stimulus
response to clozapine
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
magnesium ion binding
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
protein binding
ATP binding
kinase activity
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0