Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128148.2 | NP_001121620.1 | 4 | 505 | ||
NM_001313965.1 | NP_001300894.1 | 3 | 303 | |||
NM_003234.3 | NP_003225.2 | 4 | 646 | |||
GenBank mRNA | AB209254.1 | - | 4 | 409 | ||
AK291723.1 | - | 4 | 646 | |||
BC001188.1 | - | 4 | 419 | 3354176 | ||
BX537966.1 | - | 3 | 233 | |||
CF994006.1 | - | 4 | 533 | |||
DA460386.1 | - | 4 | 505 | |||
M11507.1 | - | 4 | 458 | |||
X01060.1 | - | 4 | 626 |