Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270483.1 | NP_001257412.1 | 2 | 564 | ||
NM_003312.5 | NP_003303.2 | 2 | 596 | |||
GenBank mRNA | AK000862.1 | - | 2 | 590 | ||
AK091874.1 | - | 1 | 1216 | |||
AK223521.1 | - | 2 | 590 | |||
AK301300.1 | - | 2 | 590 | |||
BC010148.2 | - | 2 | 573 | 4329727 | ||
BQ231617.1 | - | 2 | 589 | |||
CR456598.1 | - | 1 | 599 | |||
CR456998.1 | - | 1 | 521 | |||
D87292.1 | - | 2 | 569 |