Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256119.1 | NP_001243048.1 | 2, 3 | 378 | ||
NM_001256120.1 | NP_001243049.1 | 2, 3 | 378 | |||
NM_003491.3 | NP_003482.1 | 2, 3 | 378 | |||
GenBank mRNA | AK294081.1 | - | 2, 3 | 254 | ||
AK296593.1 | - | 2, 3 | 239 | |||
AK309083.1 | - | 2, 3 | 239 | |||
AK315997.1 | - | 2, 3 | 290 | |||
AY183134.1 | - | 2, 3 | 239 | |||
AY183135.1 | - | 2, 3 | 239 | |||
BC000308.2 | - | 2, 3 | 239 | 2820955 | ||
BC019312.1 | - | 2, 3 | 239 | 2820955 | ||
BC063377.1 | - | 2, 3 | 233 | 3050078 | ||
CR979270.1 | - | 2, 3 | 378 | |||
X77588.1 | - | 2, 3 | 223 |