Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002276.4 | NP_002267.2 | 3, 4 | 807 | ||
GenBank mRNA | AB041267.1 | - | 2, 3 | 235 | ||
AB041268.1 | - | 2, 3 | 235 | |||
AK313261.1 | - | 3, 4 | 726 | |||
BC002539.2 | - | 3, 4 | 711 | 3138504 | ||
BC007628.1 | - | 3, 4 | 711 | 3504206 | ||
BC010409.2 | - | 3, 4 | 718 | 3689178 | ||
BC067744.1 | - | 3, 4 | 726 | 30332499 | ||
BC084574.1 | - | 3, 4 | 709 | 6730698 | ||
Y00503.1 | - | 3, 4 | 697 |