Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003778.3 | NP_003769.1 | 8 | 1875 | ||
NM_212543.1 | NP_997708.1 | 9 | 1958 | |||
XM_005247855.1 | XP_005247912.1 | 8 | 1933 | |||
XM_006713798.2 | XP_006713861.1 | 9 | 2018 | |||
XM_006713799.2 | XP_006713862.1 | 9 | 2063 | |||
XM_006713800.1 | XP_006713863.1 | 8 | 1910 | |||
XM_006713801.2 | XP_006713864.1 | 10 | 2101 | |||
XM_011513260.1 | XP_011511562.1 | 10 | 2030 | |||
XM_017007401.1 | XP_016862890.1 | 7 | 1725 | |||
GenBank mRNA | AF020920.1 | - | 7 | 1764 | ||
AF038662.1 | - | 7 | 1766 | |||
AK001006.1 | - | 9 | 1931 | |||
AK123182.1 | - | 1 | 1438 | |||
AL833580.1 | - | 1 | 3956 | |||
AY359008.1 | - | 6 | 1574 | |||
BC004523.2 | - | 7 | 1624 | 3531796 | ||
BC062618.1 | - | 8 | 2097 | 5189057 | ||
BX647468.1 | - | 7 | 5406 | |||
CR749555.1 | - | 8 | 1858 |