Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002277.2 | NP_002268.2 | 7 | 1434 | ||
GenBank mRNA | BC114468.1 | - | 7 | 1404 | 40032590 | |
X86570.1 | - | 7 | 1434 |