Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003818.3 | NP_003809.1 | 10 | 1279 | ||
XM_006723660.1 | XP_006723723.1 | 10 | 1270 | |||
XM_011529390.1 | XP_011527692.1 | 11 | 1602 | |||
GenBank mRNA | AF069532.1 | - | 9 | 875 | ||
AK001414.1 | - | 10 | 1000 | |||
AK027695.1 | - | 10 | 986 | |||
AK091045.1 | - | 11 | 1602 | |||
AK095430.1 | - | 7 | 1203 | |||
AK304763.1 | - | 7 | 743 | |||
AK315489.1 | - | 10 | 1260 | |||
BC025751.1 | - | 10 | 1000 | 5199748 | ||
Y16521.1 | - | 10 | 1205 |