Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003842.4 | NP_003833.4 | 9 | 1818 | ||
NM_147187.2 | NP_671716.2 | 10 | 1731 | |||
NR_027140.1 | 9 | 1625 | ||||
XM_017013944.1 | XP_016869433.1 | 9 | 1606 | |||
GenBank mRNA | AF012535.1 | - | 10 | 1577 | ||
AF016266.1 | - | 9 | 1642 | |||
AF016268.1 | - | 10 | 1547 | |||
AF016849.1 | - | 9 | 1669 | |||
AF153687.1 | - | 10 | 1556 | |||
AF192548.1 | - | 10 | 1538 | |||
AK291216.1 | - | 9 | 1645 | |||
AK296128.1 | - | 5 | 832 | |||
AY358277.1 | - | 10 | 1577 | |||
BC001281.1 | - | 9 | 1672 | 3458466 | ||
BX537617.1 | - | 4 | 923 | |||
BX538104.1 | - | 9 | 1647 | |||
BX538179.1 | - | 4 | 923 |