Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317034.1 | NP_001303963.1 | 1 | 116 | ||
NM_001317035.1 | NP_001303964.1 | 1 | 116 | |||
NM_001317036.1 | NP_001303965.1 | 1 | 116 | |||
NM_003859.2 | NP_003850.1 | 1 | 116 | |||
NR_133648.1 | 1 | 116 | ||||
XM_011529093.2 | XP_011527395.1 | 1 | 114 | |||
GenBank mRNA | AF007875.1 | - | 1 | 54 | ||
AK289569.1 | - | 1 | 84 | |||
BC007073.1 | - | 1 | 102 | 3997453 | ||
BC008466.1 | - | 1 | 102 | 4276338 | ||
BC016322.1 | - | 1 | 87 | 4083041 | ||
BM922866.1 | - | 1 | 132 | |||
BQ441542.1 | - | 1 | 92 | |||
CR456926.1 | - | 1 | 75 | |||
D86198.1 | - | 1 | 75 | |||
DR156925.1 | - | 1 | 71 | |||
KU178444.1 | - | 1 | 75 | |||
KU178445.1 | - | 1 | 75 | |||
KU178446.1 | - | 1 | 75 |