Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317034.1 | NP_001303963.1 | 10 | 1039 | ||
NM_001317035.1 | NP_001303964.1 | 10 | 1015 | |||
NM_001317036.1 | NP_001303965.1 | 8 | 865 | |||
NM_003859.2 | NP_003850.1 | 9 | 934 | |||
NR_133648.1 | 9 | 956 | ||||
GenBank mRNA | AF007875.1 | - | 9 | 872 | ||
AK289569.1 | - | 9 | 902 | |||
BC007073.1 | - | 9 | 920 | 3997453 | ||
BC008466.1 | - | 9 | 920 | 4276338 | ||
BC016322.1 | - | 9 | 905 | 4083041 | ||
D86198.1 | - | 9 | 893 |