Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268208.1 | XP_005268265.1 | 6 | 815 | ||
XM_005268209.3 | XP_005268266.1 | 6 | 619 | |||
XM_017021765.1 | XP_016877254.1 | 6 | 618 | |||
GenBank mRNA | AB082526.1 | - | 6 | 787 | ||
AK128693.1 | - | 7 | 751 | |||
AY048580.1 | - | 6 | 681 | |||
AY080896.1 | - | 6 | 693 | |||
BC093881.1 | - | 6 | 762 | 7939726 | ||
BC112101.1 | - | 6 | 762 | 8327569 |