Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282112.1 | NP_001269041.1 | 14, 15 | 1858 | ||
NM_001282113.1 | NP_001269042.1 | 14, 15 | 1851 | |||
NM_003935.4 | NP_003926.1 | 14, 15 | 2174 | |||
XM_005261811.1 | XP_005261868.1 | 16, 17 | 2024 | |||
XM_005261813.2 | XP_005261870.1 | 15, 16 | 1889 | |||
XM_006724349.1 | XP_006724412.1 | 15, 16 | 1835 | |||
XM_006724350.1 | XP_006724413.1 | 15, 16 | 2009 | |||
XM_011530482.1 | XP_011528784.1 | 15, 16 | 2091 | |||
XM_017029038.1 | XP_016884527.1 | 14, 15 | 1837 | |||
XM_017029039.1 | XP_016884528.1 | 16, 17 | 1899 | |||
XM_017029040.1 | XP_016884529.1 | 15, 16 | 1875 | |||
XM_017029041.1 | XP_016884530.1 | 16, 17 | 1917 | |||
GenBank mRNA | AF017146.1 | - | 13, 14 | 1759 | ||
AF053082.1 | - | 13, 14 | 1759 | |||
AF125216.1 | - | 13, 14 | 1719 | |||
AK096695.1 | - | 7, 8 | 969 | |||
AK290997.1 | - | 14, 15 | 1804 | |||
AK304709.1 | - | 12, 13 | 1810 | |||
AL833505.1 | - | 15, 16 | 1888 | |||
BC002432.2 | - | 14, 15 | 2081 | 3346377 | ||
CR456596.1 | - | 13, 14 | 1673 |