Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003986.2 | NP_003977.1 | 3 | 524 | ||
XM_005253159.2 | XP_005253216.1 | 3 | 772 | |||
XM_005253160.2 | XP_005253217.1 | 3 | 769 | |||
XM_005253161.2 | XP_005253218.1 | 3 | 521 | |||
XM_011520402.1 | XP_011518704.1 | 4 | 932 | |||
XM_017018403.1 | XP_016873892.1 | 2 | 454 | |||
GenBank mRNA | AF082868.1 | - | 2 | 222 | ||
AK290108.1 | - | 2 | 269 | |||
AK313422.1 | - | 2 | 272 | |||
BC011034.1 | - | 1 | 212 | 4042355 | ||
CR456812.1 | - | 1 | 156 | |||
DA268467.1 | - | 3 | 522 |