Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004267.4 | NP_004258.2 | 2 | 2348 | ||
GenBank mRNA | AB014679.2 | - | 2 | 1657 | ||
AB014680.2 | - | 2 | 1657 | |||
AB021124.1 | - | 2 | 2793 | |||
AF083066.1 | - | 1 | 1466 | |||
AK023268.1 | - | 2 | 2298 | |||
BC017499.1 | - | 2 | 2029 | |||
BC036930.1 | - | 2 | 2259 | |||
BC068604.1 | - | 2 | 2298 | |||
BC105010.1 | - | 1 | 1428 | 8144013 | ||
BC105012.1 | - | 1 | 1428 | 8144015 | ||
EU668328.1 | - | 1 | 1428 |