Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291866.1 | NP_001278795.1 | 3 | 418 | ||
NM_004481.4 | NP_004472.1 | 3 | 340 | |||
XM_017000963.1 | XP_016856452.1 | 3 | 300 | |||
XM_017000964.1 | XP_016856453.1 | 3 | 925 | |||
XM_017000965.1 | XP_016856454.1 | 3 | 418 | |||
XM_017000966.1 | XP_016856455.1 | 3 | 243 | |||
GenBank mRNA | AK290048.1 | - | 3 | 266 | ||
AK300453.1 | - | 3 | 418 | |||
AK304029.1 | - | 2 | 170 | |||
BC041120.1 | - | 3 | 326 | 5553465 | ||
BG773173.1 | - | 3 | 346 | |||
X85019.1 | - | 3 | 254 |