Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317955.1 | NP_001304884.1 | 3, 4 | 667 | ||
NM_001317956.1 | NP_001304885.1 | 2, 3 | 801 | |||
NM_004508.3 | NP_004499.2 | 2, 3 | 684 | |||
NR_134300.1 | 1, 2 | 416 | ||||
XM_011519457.1 | XP_011517759.1 | 2, 3 | 1531 | |||
GenBank mRNA | AF271720.1 | - | 1, 2 | 169 | ||
AK222875.1 | - | 2, 3 | 204 | |||
AK303669.1 | - | 2, 3 | 438 | |||
BC005247.2 | - | 1, 2 | 182 | |||
BC006999.2 | - | 1, 2 | 182 | |||
BC019227.2 | - | 1, 2 | 182 | |||
BC022418.2 | - | 1, 2 | 182 | |||
BC025375.2 | - | 1, 2 | 182 | |||
BC057827.1 | - | 3, 4 | 655 | 30337530 | ||
BC107893.1 | - | 2, 3 | 208 | |||
BE891119.1 | - | 2, 3 | 488 | |||
BT006761.1 | - | 1, 2 | 153 | |||
BX648472.1 | - | 1, 2 | 1147 | |||
CD251816.1 | - | 2, 3 | 313 | |||
DA842794.1 | - | 2, 3 | 136 | |||
X17025.1 | - | 2, 3 | 203 |