Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317955.1 | NP_001304884.1 | 6 | 1463 | ||
NM_001317956.1 | NP_001304885.1 | 5 | 1597 | |||
NM_001317957.1 | NP_001304886.1 | 4 | 1424 | |||
NM_004508.3 | NP_004499.2 | 5 | 1480 | |||
NR_134300.1 | 4 | 1212 | ||||
NR_134301.1 | 4 | 1307 | ||||
XM_011519457.1 | XP_011517759.1 | 5 | 2327 | |||
XM_017016191.1 | XP_016871680.1 | 4 | 2154 | |||
GenBank mRNA | AK222875.1 | - | 5 | 1000 | ||
AK303669.1 | - | 5 | 1234 | |||
AK311110.1 | - | 4 | 856 | |||
BC005247.2 | - | 4 | 978 | |||
BC006999.2 | - | 4 | 978 | |||
BC019227.2 | - | 4 | 978 | |||
BC022418.2 | - | 4 | 978 | |||
BC057827.1 | - | 6 | 1451 | 30337530 | ||
BC107893.1 | - | 5 | 1004 | |||
BX537663.1 | - | 4 | 4024 | |||
BX648472.1 | - | 4 | 1943 |