Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142784.2 | NP_001136256.1 | 3 | 192 | ||
NR_052010.1 | 3 | 246 | ||||
GenBank mRNA | AK075530.1 | - | 3 | 224 | ||
AK094689.1 | - | 3 | 1499 | |||
AK292073.1 | - | 3 | 226 | |||
BC003110.1 | - | 3 | 208 | 3502059 | ||
BT009864.1 | - | 2 | 143 | |||
DA917660.1 | - | 3 | 192 | |||
DC424068.1 | - | 3 | 246 | |||
U32324.1 | - | 3 | 204 | |||
Z38102.1 | - | 2 | 148 | |||
Z46595.1 | - | 2 | 137 |