Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142784.2 | NP_001136256.1 | 5 | 421 | ||
NR_052010.1 | 5 | 475 | ||||
GenBank mRNA | AK075530.1 | - | 5 | 453 | ||
AK094689.1 | - | 5 | 1728 | |||
AK292073.1 | - | 5 | 455 | |||
BC003110.1 | - | 5 | 437 | 3502059 | ||
BT009864.1 | - | 4 | 372 | |||
DA917660.1 | - | 5 | 421 | |||
DC424068.1 | - | 5 | 475 | |||
U32324.1 | - | 5 | 433 | |||
Z38102.1 | - | 4 | 377 | |||
Z46595.1 | - | 4 | 366 |