Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323316.1 | NP_001310245.1 | 4 | 444 | ||
NM_001323318.1 | NP_001310247.1 | 2 | 281 | |||
NM_004613.3 | NP_004604.2 | 3 | 461 | |||
NM_198951.2 | NP_945189.1 | 3 | 461 | |||
XM_011529028.1 | XP_011527330.1 | 4 | 406 | |||
GenBank mRNA | AK291714.1 | - | 3 | 302 | ||
AK300292.1 | - | 2 | 122 | |||
AK314618.1 | - | 3 | 302 | |||
AY675221.1 | - | 2 | 230 | |||
BC003551.1 | - | 3 | 314 | 3544757 | ||
BX471706.1 | - | 3 | 341 | |||
DB064707.1 | - | 4 | 444 | |||
EU794665.1 | - | 3 | 330 | |||
M55153.1 | - | 3 | 365 | |||
M98478.1 | - | 3 | 362 | |||
S81734.1 | - | 3 | 362 |