Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301645.1 | NP_001288574.1 | 3 | 1013 | ||
NM_004744.4 | NP_004735.2 | 3 | 1207 | |||
GenBank mRNA | AY546085.1 | - | 2 | 1281 | ||
AY546086.1 | - | 3 | 1178 | |||
BC031053.1 | - | 3 | 1016 | 5272486 |