Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004775.3 | NP_004766.2 | 8, 9 | 1305 | ||
XM_005258387.4 | XP_005258444.1 | 8, 9 | 1070 | |||
XM_006722578.3 | XP_006722641.1 | 7, 8 | 2159 | |||
XM_006722579.3 | XP_006722642.1 | 9, 10 | 1018 | |||
XM_017026088.1 | XP_016881577.1 | 7, 8 | 2156 | |||
XM_017026089.1 | XP_016881578.1 | 8, 9 | 1476 | |||
XM_017026090.1 | XP_016881579.1 | 6, 7 | 2919 | |||
XM_017026091.1 | XP_016881580.1 | 6, 7 | 773 | |||
XM_017026092.1 | XP_016881581.1 | 7, 8 | 1351 | |||
GenBank mRNA | AB024742.2 | - | 8, 9 | 1014 | ||
AB209194.1 | - | 4, 5 | 638 | |||
AF038664.1 | - | 8, 9 | 1283 | |||
AF069054.1 | - | 7, 8 | 1036 | |||
AF097159.1 | - | 8, 9 | 1305 | |||
AK314860.1 | - | 8, 9 | 1163 | |||
BC069620.1 | - | 7, 8 | 969 | 7262397 | ||
BC069642.1 | - | 8, 9 | 1085 | 7262468 | ||
BC074835.2 | - | 8, 9 | 1026 | 30915327 | ||
BC074884.2 | - | 8, 9 | 1026 | 30915473 |