Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004792.2 | NP_004783.2 | 5 | 377 | ||
XM_005246966.1 | XP_005247023.1 | 5 | 334 | |||
XM_005246967.1 | XP_005247024.1 | 5 | 381 | |||
XM_017005302.1 | XP_016860791.1 | 3 | 173 | |||
GenBank mRNA | AK307552.1 | - | 2 | 168 | ||
BC001555.1 | - | 6 | 337 | |||
BC063711.1 | - | 5 | 479 | |||
BC073875.1 | - | 4 | 232 | |||
BC109373.1 | - | 5 | 381 | |||
BC111693.1 | - | 5 | 364 | 40034815 | ||
U40763.1 | - | 6 | 642 | |||
X99717.1 | - | 5 | 314 |