Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004792.2 | NP_004783.2 | 12 | 1195 | ||
XM_005246966.1 | XP_005247023.1 | 12 | 1152 | |||
XM_005246967.1 | XP_005247024.1 | 12 | 1199 | |||
GenBank mRNA | AK307552.1 | - | 9 | 977 | ||
BC001555.1 | - | 13 | 1155 | |||
BC073875.1 | - | 11 | 1041 | |||
BC109373.1 | - | 12 | 1199 | |||
BC111693.1 | - | 11 | 1137 | 40034815 | ||
U40763.1 | - | 13 | 1460 | |||
X99717.1 | - | 12 | 1132 |