Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145453.2 | NP_001138925.1 | 6 | 1549 | ||
NM_005264.5 | NP_005255.1 | 7 | 1564 | |||
NM_145793.3 | NP_665736.1 | 5 | 1098 | |||
XM_005269696.1 | XP_005269753.1 | 7 | 1174 | |||
XM_011539634.1 | XP_011537936.1 | 7 | 1139 | |||
GenBank mRNA | AF038421.1 | - | 7 | 1391 | ||
AF042080.1 | - | 6 | 945 | |||
AK292886.1 | - | 6 | 1110 | |||
AK302914.1 | - | 3 | 564 | |||
BC014962.1 | - | 5 | 1073 | 4874042 | ||
U95847.1 | - | 5 | 1064 | |||
U97144.1 | - | 5 | 1081 |