Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252049.1 | NP_001238978.1 | 5 | 586 | ||
NM_001252050.1 | NP_001238979.1 | 4 | 481 | |||
NM_001252051.1 | NP_001238980.1 | 4 | 481 | |||
NM_001252052.1 | NP_001238981.1 | 4 | 481 | |||
NM_001252053.1 | NP_001238982.1 | 5 | 586 | |||
NM_005389.2 | NP_005380.2 | 5 | 586 | |||
XM_011535868.2 | XP_011534170.1 | 5 | 787 | |||
GenBank mRNA | AA442100.1 | - | 3 | 203 | ||
AK098739.1 | - | 4 | 464 | |||
AK129778.1 | - | 5 | 502 | |||
AK289724.1 | - | 5 | 554 | |||
AK304532.1 | - | 4 | 402 | |||
AL539512.3 | - | 4 | 453 | |||
BC007501.2 | - | 5 | 528 | 3010742 | ||
BC008748.2 | - | 5 | 464 | 3346722 | ||
BC018569.2 | - | 1 | 71 | |||
BE262147.1 | - | 4 | 366 | |||
CX786396.1 | - | 5 | 472 | |||
D13892.1 | - | 5 | 529 | |||
D25545.1 | - | 5 | 452 | |||
D25546.1 | - | 5 | 452 | |||
D25547.1 | - | 5 | 452 | |||
DC356391.1 | - | 5 | 569 | |||
DC421281.1 | - | 5 | 576 | |||
M93008.1 | - | 5 | 416 | |||
M93009.1 | - | 1 | 22 |