Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252049.1 | NP_001238978.1 | 8 | 1626 | ||
NM_001252050.1 | NP_001238979.1 | 7 | 1568 | |||
NM_001252051.1 | NP_001238980.1 | 7 | 1568 | |||
NM_001252052.1 | NP_001238981.1 | 7 | 1521 | |||
NM_005389.2 | NP_005380.2 | 8 | 1673 | |||
XM_011535868.2 | XP_011534170.1 | 8 | 1827 | |||
GenBank mRNA | AK098739.1 | - | 7 | 1551 | ||
AK129778.1 | - | 8 | 1589 | |||
BC007501.2 | - | 8 | 1568 | 3010742 | ||
BC018569.2 | - | 4 | 1158 | |||
D13892.1 | - | 8 | 1569 | |||
D25545.1 | - | 8 | 1539 | |||
D25546.1 | - | 8 | 1492 | |||
M93009.1 | - | 4 | 1109 |