Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166356.1 | NP_001159828.1 | 3 | 508 | ||
NM_001166357.1 | NP_001159829.1 | 3 | 570 | |||
NM_001166358.1 | NP_001159830.1 | 3 | 469 | |||
NM_001166359.1 | NP_001159831.1 | 3 | 362 | |||
NM_005412.5 | NP_005403.2 | 3 | 508 | |||
XM_011538675.2 | XP_011536977.1 | 3 | 420 | |||
XM_011538676.2 | XP_011536978.1 | 3 | 411 | |||
GenBank mRNA | AK223555.1 | - | 3 | 370 | ||
AK296183.1 | - | 3 | 322 | |||
AK303827.1 | - | 3 | 370 | |||
AK315916.1 | - | 3 | 562 | |||
BC011911.2 | - | 3 | 312 | 4139001 | ||
BC013677.2 | - | 3 | 372 | 3863368 | ||
BC032584.1 | - | 3 | 381 | 5499481 | ||
BC044211.1 | - | 3 | 348 | 5521082 | ||
BT006866.1 | - | 3 | 302 | |||
BX647711.1 | - | 3 | 429 | |||
CN395976.1 | - | 3 | 364 | |||
DA052422.1 | - | 3 | 474 | |||
EU668344.1 | - | 3 | 312 | |||
KU178333.1 | - | 3 | 302 | |||
L11932.1 | - | 2 | 213 |