Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002143.2 | NP_002134.2 | 2, 3 | 426 | ||
XM_005270792.2 | XP_005270849.1 | 2, 3 | 562 | |||
XM_017001118.1 | XP_016856607.1 | 2, 3 | 898 | |||
GenBank mRNA | AK313907.1 | - | 2, 3 | 430 | ||
BC001777.1 | - | 2, 3 | 426 | 3355354 | ||
BT019750.1 | - | 1, 2 | 386 | |||
BT019789.1 | - | 1, 2 | 386 | |||
D16593.1 | - | 1, 2 | 396 |