Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005420.2 | NP_005411.1 | 6, 7 | 703 | ||
GenBank mRNA | AK312236.1 | - | 5, 6 | 602 | ||
BC027956.1 | - | 6, 7 | 703 | 5210178 | ||
CR407621.1 | - | 5, 6 | 590 | |||
S77383.1 | - | 6, 7 | 626 | |||
U08098.1 | - | 6, 7 | 696 | |||
U55764.1 | - | 1, 2 | 49 | |||
Y11195.1 | - | 6, 7 | 652 |