Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003931.3 | NP_001003931.3 | 2 | 543 | ||
NM_005485.5 | NP_005476.4 | 2 | 548 | |||
XM_005264779.4 | XP_005264836.2 | 3 | 688 | |||
XM_017005490.1 | XP_016860979.1 | 3 | 675 | |||
GenBank mRNA | AB208803.1 | - | 2 | 428 | ||
AF083068.1 | - | 2 | 267 | |||
AY126341.1 | - | 2 | 378 | |||
BC014260.2 | - | 1 | 149 | 4763951 | ||
BG818516.1 | - | 3 | 627 | |||
BI554046.1 | - | 2 | 482 | |||
Y16836.1 | - | 2 | 502 |