Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003931.3 | NP_001003931.3 | 7, 8 | 1425 | ||
NM_005485.5 | NP_005476.4 | 7, 8 | 1430 | |||
XM_005264779.4 | XP_005264836.2 | 8, 9 | 1570 | |||
XM_017005490.1 | XP_016860979.1 | 8, 9 | 1557 | |||
GenBank mRNA | AB208803.1 | - | 7, 8 | 1310 | ||
AF083068.1 | - | 7, 8 | 1149 | |||
AL050034.1 | - | 5, 6 | 798 | |||
AY126341.1 | - | 7, 8 | 1260 | |||
BC014260.2 | - | 6, 7 | 1031 | 4763951 | ||
Y16836.1 | - | 7, 8 | 1384 |