Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258357.1 | NP_001245286.1 | 4 | 627 | ||
NM_001258358.1 | NP_001245287.1 | 4 | 639 | |||
NM_001258359.1 | NP_001245288.1 | 4 | 1033 | |||
NM_002149.3 | NP_002140.2 | 4 | 853 | |||
NM_134421.2 | NP_602293.1 | 5 | 995 | |||
XM_005246160.1 | XP_005246217.1 | 5 | 918 | |||
XM_005246161.1 | XP_005246218.1 | 5 | 1041 | |||
XM_005246162.1 | XP_005246219.1 | 5 | 1044 | |||
XM_005246163.1 | XP_005246220.1 | 6 | 1091 | |||
XM_011510346.2 | XP_011508648.1 | 4 | 697 | |||
XM_011510347.1 | XP_011508649.1 | 6 | 1138 | |||
XM_011510348.1 | XP_011508650.1 | 5 | 931 | |||
XM_011510349.1 | XP_011508651.1 | 4 | 868 | |||
XM_017003950.1 | XP_016859439.1 | 6 | 2585 | |||
XM_017003951.1 | XP_016859440.1 | 6 | 1068 | |||
XM_017003952.1 | XP_016859441.1 | 4 | 813 | |||
XM_017003953.1 | XP_016859442.1 | 5 | 948 | |||
GenBank mRNA | AF070616.1 | - | 2 | 222 | ||
AK000596.1 | - | 5 | 1001 | |||
AK292280.1 | - | 4 | 1033 | |||
BC009846.2 | - | 4 | 828 | 4100091 | ||
BC017028.1 | - | 4 | 820 | 3902811 | ||
BC017482.1 | - | 4 | 843 | 3504942 | ||
BI601950.1 | - | 4 | 644 | |||
BT009835.1 | - | 2 | 469 | |||
BU145561.1 | - | 4 | 634 | |||
CR541975.1 | - | 2 | 469 | |||
CR749837.1 | - | 6 | 968 | |||
D16227.1 | - | 3 | 616 |