Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12RB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290023.1 | NP_001276952.1 | 3 | 335 | ||
NM_001290024.1 | NP_001276953.1 | 4 | 326 | |||
NM_005535.2 | NP_005526.1 | 3 | 335 | |||
NM_153701.2 | NP_714912.1 | 3 | 335 | |||
XM_006722741.3 | XP_006722804.2 | 4 | 528 | |||
XM_011527966.2 | XP_011526268.1 | 4 | 549 | |||
XM_011527967.2 | XP_011526269.1 | 4 | 535 | |||
XM_011527968.2 | XP_011526270.1 | 4 | 543 | |||
XM_011527969.2 | XP_011526271.1 | 4 | 529 | |||
XM_011527970.2 | XP_011526272.1 | 4 | 548 | |||
XM_011527971.2 | XP_011526273.1 | 4 | 546 | |||
XM_011527972.2 | XP_011526274.1 | 4 | 546 | |||
XM_011527973.2 | XP_011526275.1 | 3 | 340 | |||
XM_011527974.2 | XP_011526276.1 | 3 | 328 | |||
XM_011527975.2 | XP_011526277.1 | 4 | 526 | |||
XM_011527976.2 | XP_011526278.1 | 4 | 546 | |||
XM_011527977.2 | XP_011526279.1 | 4 | 519 | |||
GenBank mRNA | AK225919.1 | - | 3 | 266 | ||
AK290423.1 | - | 3 | 263 | |||
BC029121.1 | - | 3 | 224 | |||
BC137404.1 | - | 3 | 180 | 9021402 | ||
BC137406.1 | - | 3 | 180 | 9021404 | ||
BX647221.1 | - | 4 | 326 | |||
DA008204.1 | - | 3 | 335 | |||
U03187.1 | - | 3 | 219 |