Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005729.3 | NP_005720.1 | 6 | 1855 | ||
GenBank mRNA | BC013204.1 | - | 6 | 1832 | ||
BC110299.1 | - | 5 | 1756 | 5215228 |