Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282704.1 | NP_001269633.1 | 5 | 689 | ||
NM_001282705.1 | NP_001269634.1 | 4 | 711 | |||
NM_001282706.1 | NP_001269635.1 | 4 | 541 | |||
NM_001282707.1 | NP_001269636.1 | 4 | 516 | |||
NM_005742.3 | NP_005733.1 | 3 | 338 | |||
XM_011510308.1 | XP_011508610.1 | 3 | 279 | |||
GenBank mRNA | AK127433.1 | - | 5 | 689 | ||
AK131234.1 | - | 4 | 707 | |||
AK225496.1 | - | 2 | 84 | |||
AK289428.1 | - | 3 | 214 | |||
AK294347.1 | - | 4 | 431 | |||
BC001312.1 | - | 3 | 294 | 3454007 | ||
BF979486.1 | - | 343 | ||||
D49489.1 | - | 289 | ||||
DA976962.1 | - | 3 | 338 | |||
HY166449.1 | - | 3 | 328 | |||
U79278.1 | - | 2 | 140 |