Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005894.2 | NP_005885.1 | 6 | 1824 | ||
GenBank mRNA | AF147312.1 | - | 1 | 275 | ||
BC033586.1 | - | 6 | 1712 | 5225975 | ||
U82812.1 | - | 6 | 1740 |