Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005911.5 | NP_005902.1 | 5 | 795 | ||
GenBank mRNA | AK291126.1 | - | 5 | 603 | ||
AK293841.1 | - | 5 | 605 | |||
AK297758.1 | - | 5 | 498 | |||
AK308591.1 | - | 4 | 581 | |||
AK308758.1 | - | 5 | 463 | |||
AK309338.1 | - | 5 | 603 | |||
AK316411.1 | - | 5 | 753 | |||
BC001686.2 | - | 5 | 592 | 3010820 | ||
BC001854.2 | - | 5 | 592 | 3010820 | ||
BI549760.1 | - | 5 | 614 | |||
DQ083239.1 | - | 5 | 531 | |||
X68836.1 | - | 5 | 550 |