Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006011.3 | NP_006002.1 | 6 | 1138 | ||
XM_017022642.1 | XP_016878131.1 | 6 | 1301 | |||
XM_017022643.1 | XP_016878132.1 | 5 | 1074 | |||
GenBank mRNA | AK313924.1 | - | 6 | 1086 | ||
BC069584.1 | - | 6 | 1007 | |||
BC096202.3 | - | 6 | 1007 | 40004644 | ||
BC096203.3 | - | 6 | 1007 | 40004645 | ||
BC096204.3 | - | 6 | 1007 | 40004646 | ||
BC096205.3 | - | 5 | 944 | 40004648 | ||
L29556.1 | - | 3 | 524 | |||
U33551.1 | - | 6 | 983 | |||
U82762.1 | - | 6 | 1008 |