Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002161.5 | NP_002152.2 | 6 | 669 | ||
NM_013417.3 | NP_038203.2 | 6 | 839 | |||
NR_073446.1 | 5 | 512 | ||||
GenBank mRNA | AB209234.1 | - | 6 | 578 | ||
AK293014.1 | - | 6 | 596 | |||
BC065552.1 | - | 5 | 511 | 6043344 | ||
BX537417.1 | - | 6 | 607 | |||
BX537429.1 | - | 6 | 609 | |||
BX538109.1 | - | 5 | 452 | |||
BX647071.1 | - | 6 | 609 | |||
BX647236.1 | - | 6 | 601 | |||
BX647430.1 | - | 6 | 603 | |||
BX648567.1 | - | 6 | 609 | |||
D28473.1 | - | 6 | 757 | |||
U04953.1 | - | 6 | 771 |