Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006223.3 | NP_006214.2 | 4 | 978 | ||
NR_033187.1 | 3 | 904 | ||||
GenBank mRNA | AB009690.1 | - | 4 | 899 | ||
AF143096.1 | - | 4 | 892 | |||
BC005234.1 | - | 4 | 897 | |||
BC070288.1 | - | 4 | 897 | |||
BC111394.1 | - | 4 | 898 |