Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319075.1 | NP_001306004.1 | 2 | 1166 | ||
NM_006577.5 | NP_006568.2 | 2 | 1300 | |||
GenBank mRNA | AB049584.1 | - | 2 | 1295 | ||
AF092051.2 | - | 2 | 1297 | |||
AF288208.1 | - | 2 | 1290 | |||
AF288209.1 | - | 1 | 1092 | |||
AJ006077.1 | - | 1 | 1065 | |||
AK002009.1 | - | 1 | 995 | |||
BC030579.2 | - | 2 | 1169 | 4828158 | ||
BC047933.1 | - | 2 | 1300 | 5767816 |